missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-methoxybenzoate, 98+%

$58.81 - $671.12

Chemical Identifiers

CAS 94-30-4
Molecular Formula C10H12O3
Formula Weight 180.2
MDL Number MFCD00015151
Beilstein 2209700
Synonym Ethyl p-anisate; 4-Methoxybenzoic acid ethyl ester
Chemical Name or Material Ethyl 4-methoxybenzoate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA140080B SDS Alfa Aesar™
A140080B
1000g Each for $671.12
AAA1400818 SDS Alfa Aesar™
A1400818
50g Each for $58.81
AAA1400830 SDS Alfa Aesar™
A1400830
250g Each for $195.49
Specifications Description & Specifications

Chemical Identifiers

94-30-4
180.2
2209700
Ethyl 4-methoxybenzoate
C10H12O3
MFCD00015151
Ethyl p-anisate; 4-Methoxybenzoic acid ethyl ester

Specifications

94-30-4
180.2
2209700
263°C
1000g
Ethyl 4-methoxybenzoate
C10H12O3
MFCD00015151
Ethyl p-anisate; 4-Methoxybenzoic acid ethyl ester
7° to 8°C
138°C (280°F)
≥98%

Safety and Handling

RTECSNumber : BZ4697000

TSCA : TSCA