Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-isopropylpiperazine-1-carboxylate, 98%

$42.19 - $161.62

Chemical Identifiers

Chemical Name or Material Ethyl 4-isopropylpiperazine-1-carboxylate
CAS 61014-91-3
Molecular Formula C10H20N2O2
Formula Weight 200.28
MDL Number MFCD08064222
Synonym Ethyl 4-(1-methylethyl)piperazine-1-carboxylate; 4-Isopropylpiperazine-1-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2534103

 
alfa aesar™
B2534103
1g Each for $42.19

AAB2534106

 
alfa aesar™
B2534106
5g Each for $161.62
Description & Specifications

Specifications

Formula Weight 200.28
Chemical Name or Material Ethyl 4-isopropylpiperazine-1-carboxylate
Assay 98%
Molecular Formula C10H20N2O2
MDL Number MFCD08064222
CAS 61014-91-3
Synonym Ethyl 4-(1-methylethyl)piperazine-1-carboxylate; 4-Isopropylpiperazine-1-carboxylic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 128° to 130°C (14mmHg)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.