Please login to your online account to display your discounted pricing

Ethyl 4-isocyanatobenzoate, 99+%, ACROS Organics™


Quantity 25g
Packaging Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  170310250

 View more versions of this product

Catalog No. AC170310250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid After Melting
Assay ≥99.0%
Formula Weight 191.19
Quantity 25g
Color Clear colorless to yellow
Packaging Glass Bottle in Tin Can
EINECS Number 250-344-2
Chemical Name or Material Ethyl 4-isocyanatobenzoate
Name Note 99+%
Storage Information Product may develop some turbidity or turbidity
Assay Percent Range 99+%
Molecular Formula C10H9NO3
Linear Formula OCNC6H4CO2C2H5
MDL Number MFCD00002027
CAS 30806-83-8
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Ethyl 4-isocyanatobenzoate
Name Note 99+%
CAS 30806-83-8
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C10H9NO3
Linear Formula OCNC6H4CO2C2H5
Formula Weight 191.19
MDL Number MFCD00002027
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.

SDS & Safety Documents