missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-iodobenzoate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 249640250

 View more versions of this product

Catalog No. AC249640250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Ethyl 4-iodobenzoate
Authentic
1.5865 to 1.5885
281.0°C to 283.0°C
25mL
51934-41-9
C9H9IO2
MFCD00017344
Solubility in water: insoluble. Other solubilities: insoluble in non-polar solvents,soluble in polar solvents
CCOC(=O)C1=CC=C(C=C1)I
276.05
276.05
97%
1.6500g/mL
Glass bottle
1.65
Colorless to Yellow
97%
96% min. (GC)
IC6H4CO2CH2CH3
4-iodo-benzoic acid ethyl ester, 4-iodobenzoic acid ethyl ester, benzoic acid, 4-iodo-, ethyl ester, benzoicacid,4-iodo-,ethylester, ethyl p-iodobenzoate, ethyl,4-iodobenzoate, ethyl-4 iodobenzoate, ethyl-4-iodobenzoate, ethyl4-iodobenzoate, p-iodobenzoic acid ethyl ester
YCBJOQUNPLTBGG-UHFFFAOYSA-N
ethyl 4-iodobenzoate
142891
Liquid

Chemical Identifiers

51934-41-9
276.05
YCBJOQUNPLTBGG-UHFFFAOYSA-N
142891
CCOC(=O)C1=CC=C(C=C1)I
C9H9IO2
MFCD00017344
4-iodo-benzoic acid ethyl ester, 4-iodobenzoic acid ethyl ester, benzoic acid, 4-iodo-, ethyl ester, benzoicacid,4-iodo-,ethylester, ethyl p-iodobenzoate, ethyl,4-iodobenzoate, ethyl-4 iodobenzoate, ethyl-4-iodobenzoate, ethyl4-iodobenzoate, p-iodobenzoic acid ethyl ester
ethyl 4-iodobenzoate
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents