Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans, 97%

$77.46 - $238.07

Chemical Identifiers

Chemical Name or Material Ethyl 4-hydroxycyclohexanecarboxylate
CAS 17159-80-7
Name Note cis + trans
Molecular Formula C9H16O3
Formula Weight 172.22
MDL Number MFCD00192156
Synonym 4-Hydroxycyclohexanecarboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2557206

  SDS
alfa aesar™
B2557206
5g Each for $77.46

AAB2557214

  SDS
alfa aesar™
B2557214
25g Each for $238.07
Description & Specifications

Specifications

Formula Weight 172.22
MDL Number MFCD00192156
Molecular Formula C9H16O3
Chemical Name or Material Ethyl 4-hydroxycyclohexanecarboxylate
Name Note cis + trans
Assay 97%
Boiling Point 127° to 134°C (0.1mmHg)
CAS 17159-80-7
Synonym 4-Hydroxycyclohexanecarboxylic acid ethyl ester
Quantity 5g
Flash Point >110°C (230°F)

Applications
Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans is used in the synthesis of other chemicals and as pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.