missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2557214
Description
Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans is used in the synthesis of other chemicals and as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 4-hydroxycyclohexanecarboxylate, cis + trans is used in the synthesis of other chemicals and as pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
Ethyl 4-hydroxycyclohexanecarboxylate | |
17159-80-7 | |
127°C to 134°C (0.1 mmHg) | |
C9H16O3 | |
MFCD00192156,MFCD16295187 | |
ethyl 4-hydroxycyclohexanecarboxylate, trans-ethyl 4-hydroxycyclohexanecarboxylate, ethyl trans-4-hydroxycyclohexanecarboxylate, 4-hydroxy-cyclohexanecarboxylic acid ethyl ester, ethyl 4-hydroxycyclohexane carboxylate, cis-ethyl 4-hydroxycyclohexanecarboxylate, cyclohexanecarboxylic acid, 4-hydroxy-, ethyl ester, 4-hydroxycyclohexanecarboxylic acid ethyl ester, trans-4-hydroxycyclohexanecarboxylic acid ethyl ester, cyclohexanecarboxylic acid, 4-hydroxy-, ethyl ester, cis | |
BZKQJSLASWRDNE-UHFFFAOYSA-N | |
ethyl 4-hydroxycyclohexane-1-carboxylate | |
86973 | |
97% |
cis + trans | |
1.068 | |
>110(230°F) | |
1.467 | |
25 g | |
Not miscible or difficult to mix in water. | |
CCOC(=O)C1CCC(O)CC1 | |
172.22 | |
172.22 |
Chemical Identifiers
17159-80-7 | |
172.22 | |
BZKQJSLASWRDNE-UHFFFAOYSA-N | |
86973 | |
CCOC(=O)C1CCC(O)CC1 |
C9H16O3 | |
MFCD00192156,MFCD16295187 | |
ethyl 4-hydroxycyclohexanecarboxylate, trans-ethyl 4-hydroxycyclohexanecarboxylate, ethyl trans-4-hydroxycyclohexanecarboxylate, 4-hydroxy-cyclohexanecarboxylic acid ethyl ester, ethyl 4-hydroxycyclohexane carboxylate, cis-ethyl 4-hydroxycyclohexanecarboxylate, cyclohexanecarboxylic acid, 4-hydroxy-, ethyl ester, 4-hydroxycyclohexanecarboxylic acid ethyl ester, trans-4-hydroxycyclohexanecarboxylic acid ethyl ester, cyclohexanecarboxylic acid, 4-hydroxy-, ethyl ester, cis | |
ethyl 4-hydroxycyclohexane-1-carboxylate |
Safety and Handling
EINECSNumber : 241-215-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only