Please login to your online account to display your discounted pricing

Ethyl-4-hydroxy-6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylate, 95%, Maybridge


Quantity 25g

Manufacturer: fisher scientific company  BTB12129EB

 View more versions of this product

Catalog No. BTB12129EB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 197.19
Quantity 25g
Assay 95%
CAS Max % 100.0
Molecular Formula C9H11NO4
MDL Number MFCD02929397
Chemical Name or Material Ethyl 2, 4-dihydroxy-6-methylnicotinate
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
CAS Min % 94.5
Synonym Ethyl-4-hydroxy-6-methyl-2-oxo-1, 2-dihydropyridine-3-carboxylate (IUPAC)
Health Hazard 1 Warning
CAS 10350-10-4
Assay Percent Range 95%

Specifications

Chemical Name or Material Ethyl 2, 4-dihydroxy-6-methylnicotinate
CAS 10350-10-4
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H11NO4
Formula Weight 197.19
MDL Number MFCD02929397
Synonym Ethyl-4-hydroxy-6-methyl-2-oxo-1, 2-dihydropyridine-3-carboxylate (IUPAC)
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

SDS & Safety Documents