missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-hydroxy-2-methylpyrimidine-5-carboxylate, 97%

$68.31 - $309.96

Chemical Identifiers

CAS 53135-24-3
Molecular Formula C8H10N2O3
Molecular Weight (g/mol) 182.179
MDL Number MFCD00223606
InChI Key KTZQDIINDVWLES-UHFFFAOYSA-N
Synonym ethyl 4-hydroxy-2-methylpyrimidine-5-carboxylate, ethyl 2-methyl-6-oxo-1,6-dihydropyrimidine-5-carboxylate, 4-hydroxy-2-methyl-pyrimidine-5-carboxylic acid ethyl ester, 4-hydroxy-2-methylpyrimidine-5-carboxylic acidethyl ester, ethyl4-hydroxy-2-methylpyrimidine-5-carboxylate, ethyl 4-hydroxy-2-methyl-pyrimidine-5-carboxylate, 4-hydroxy-2-methylpyrimidine-5-carboxylic acid ethyl ester, ethyl 4-hydroxy-2-methyl-5-pyrimidine carboxylate, 4-hydroxy-2-methyl-pyrimidine-5-carboxylic acidethyl ester, 5-pyrimidinecarboxylic acid, 4-hydroxy-2-methyl-, ethyl ester
PubChem CID 293750
IUPAC Name ethyl 2-methyl-6-oxo-1H-pyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(NC1=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6545303 View Documents Alfa Aesar™
H6545303
1g Each for $68.31
 
AAH6545306 View Documents Alfa Aesar™
H6545306
5g Each for $309.96
 
Specifications

Chemical Identifiers

53135-24-3
182.179
KTZQDIINDVWLES-UHFFFAOYSA-N
293750
CCOC(=O)C1=CN=C(NC1=O)C
C8H10N2O3
MFCD00223606
ethyl 4-hydroxy-2-methylpyrimidine-5-carboxylate, ethyl 2-methyl-6-oxo-1,6-dihydropyrimidine-5-carboxylate, 4-hydroxy-2-methyl-pyrimidine-5-carboxylic acid ethyl ester, 4-hydroxy-2-methylpyrimidine-5-carboxylic acidethyl ester, ethyl4-hydroxy-2-methylpyrimidine-5-carboxylate, ethyl 4-hydroxy-2-methyl-pyrimidine-5-carboxylate, 4-hydroxy-2-methylpyrimidine-5-carboxylic acid ethyl ester, ethyl 4-hydroxy-2-methyl-5-pyrimidine carboxylate, 4-hydroxy-2-methyl-pyrimidine-5-carboxylic acidethyl ester, 5-pyrimidinecarboxylic acid, 4-hydroxy-2-methyl-, ethyl ester
ethyl 2-methyl-6-oxo-1H-pyrimidine-5-carboxylate

Specifications

1g
C8H10N2O3
152549
KTZQDIINDVWLES-UHFFFAOYSA-N
ethyl 2-methyl-6-oxo-1H-pyrimidine-5-carboxylate
293750
Solid
Ethyl 4-hydroxy-2-methylpyrimidine-5-carboxylate
53135-24-3
MFCD00223606
ethyl 4-hydroxy-2-methylpyrimidine-5-carboxylate, ethyl 2-methyl-6-oxo-1,6-dihydropyrimidine-5-carboxylate, 4-hydroxy-2-methyl-pyrimidine-5-carboxylic acid ethyl ester, 4-hydroxy-2-methylpyrimidine-5-carboxylic acidethyl ester, ethyl4-hydroxy-2-methylpyrimidine-5-carboxylate, ethyl 4-hydroxy-2-methyl-pyrimidine-5-carboxylate, 4-hydroxy-2-methylpyrimidine-5-carboxylic acid ethyl ester, ethyl 4-hydroxy-2-methyl-5-pyrimidine carboxylate, 4-hydroxy-2-methyl-pyrimidine-5-carboxylic acidethyl ester, 5-pyrimidinecarboxylic acid, 4-hydroxy-2-methyl-, ethyl ester
CCOC(=O)C1=CN=C(NC1=O)C
182.179
182.18
97%