Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl (4-formyl-2-methoxyphenoxy)acetate

$116.99 - $404.82

Chemical Identifiers

Chemical Name or Material Ethyl (4-formyl-2-methoxyphenoxy)acetate
CAS 51264-70-1
Molecular Formula C12H14O5
Formula Weight 238.24
MDL Number MFCD01764358
Synonym (4-Formyl-2-methoxyphenoxy)acetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5049103

 
alfa aesar™
H5049103
1g Each for $404.82

AAH50491MD

 
alfa aesar™
H50491MD
250mg Each for $116.99
Description & Specifications

Specifications

Formula Weight 238.24
MDL Number MFCD01764358
CAS 51264-70-1
Synonym (4-Formyl-2-methoxyphenoxy)acetic acid ethyl ester
Chemical Name or Material Ethyl (4-formyl-2-methoxyphenoxy)acetate
Molecular Formula C12H14O5
Quantity 1g