missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl (4-fluorobenzoyl)acetate, 95%

$30.53 - $342.35

Chemical Identifiers

CAS 1999-00-4
Molecular Formula C11H11FO3
Molecular Weight (g/mol) 210.204
MDL Number MFCD03093631
InChI Key SJUXLKYJKQBZLM-UHFFFAOYSA-N
Synonym ethyl 3-4-fluorophenyl-3-oxopropanoate, ethyl 4-fluorobenzoylacetate, ethyl 4-fluorobenzoyl acetate, 4-fluorobenzoylacetic acid ethyl ester, ethyl p-fluorobenzoyl acetate, 4-fluorobenzoyl acetic acid ethyl ester, ethyl 3-4-fluorophenyl-3-oxopropionate, ethyl 3-4'-fluorophenyl-3-oxopropanoate, ethyl 4'-fluorobenzoylacetate, 3-4-fluoro-phenyl-3-oxo-propionic acid ethyl ester
PubChem CID 2758844
IUPAC Name ethyl 3-(4-fluorophenyl)-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1=CC=C(C=C1)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61813MD View Documents Alfa Aesar™
H61813MD
250mg Each for $30.53
 
AAH6181303 View Documents Alfa Aesar™
H6181303
1g Each for $89.91
 
AAH6181306 View Documents Alfa Aesar™
H6181306
5g Each for $342.35
 
Specifications

Chemical Identifiers

1999-00-4
210.204
SJUXLKYJKQBZLM-UHFFFAOYSA-N
2758844
CCOC(=O)CC(=O)C1=CC=C(C=C1)F
C11H11FO3
MFCD03093631
ethyl 3-4-fluorophenyl-3-oxopropanoate, ethyl 4-fluorobenzoylacetate, ethyl 4-fluorobenzoyl acetate, 4-fluorobenzoylacetic acid ethyl ester, ethyl p-fluorobenzoyl acetate, 4-fluorobenzoyl acetic acid ethyl ester, ethyl 3-4-fluorophenyl-3-oxopropionate, ethyl 3-4'-fluorophenyl-3-oxopropanoate, ethyl 4'-fluorobenzoylacetate, 3-4-fluoro-phenyl-3-oxo-propionic acid ethyl ester
ethyl 3-(4-fluorophenyl)-3-oxopropanoate

Specifications

117°C to 120°C
1999-00-4
MFCD03093631
SJUXLKYJKQBZLM-UHFFFAOYSA-N
ethyl 3-(4-fluorophenyl)-3-oxopropanoate
2758844
95%
250mg
C11H11FO3
ethyl 3-4-fluorophenyl-3-oxopropanoate, ethyl 4-fluorobenzoylacetate, ethyl 4-fluorobenzoyl acetate, 4-fluorobenzoylacetic acid ethyl ester, ethyl p-fluorobenzoyl acetate, 4-fluorobenzoyl acetic acid ethyl ester, ethyl 3-4-fluorophenyl-3-oxopropionate, ethyl 3-4'-fluorophenyl-3-oxopropanoate, ethyl 4'-fluorobenzoylacetate, 3-4-fluoro-phenyl-3-oxo-propionic acid ethyl ester
CCOC(=O)CC(=O)C1=CC=C(C=C1)F
210.204
210.2
Ethyl (4-fluorobenzoyl)acetate