Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-fluorobenzoate, 99%

$30.47 - $272.45

Chemical Identifiers

Chemical Name or Material Ethyl 4-fluorobenzoate
CAS 451-46-7
Molecular Formula C9H9FO2
Formula Weight 168.17
MDL Number MFCD00000351
Beilstein 1866794
Merck Index 14,4171
Synonym 4-Fluorobenzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2148406

 
alfa aesar™
B2148406
5g Each for $30.47

AAB2148414

 
alfa aesar™
B2148414
25g Each for $90.05

AAB2148422

 
alfa aesar™
B2148422
100g Each for $272.45
Description & Specifications

Specifications

Formula Weight 168.17
Chemical Name or Material Ethyl 4-fluorobenzoate
TSCA TSCA
Odor Recognizable
Assay 99%
Molecular Formula C9H9FO2
MDL Number MFCD00000351
CAS 451-46-7
Beilstein 1866794
Merck Index 14,4171
Synonym 4-Fluorobenzoic acid ethyl ester
Boiling Point 209° to 210°C
Melting Point 25° to 27°C
Quantity 5g
Flash Point 81°C (177°F)