Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-ethoxyphenylacetate, 97+%

$62.96 - $227.81

Chemical Identifiers

Chemical Name or Material Ethyl 4-ethoxyphenylacetate
CAS 40784-88-1
Molecular Formula C12H16O3
Formula Weight 208.26
MDL Number MFCD00051917
Beilstein 2969074
Synonym 4-Ethoxyphenylacetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2126806

  SDS
alfa aesar™
B2126806
5g Each for $62.96

AAB2126814

  SDS
alfa aesar™
B2126814
25g Each for $227.81
Description & Specifications

Specifications

Formula Weight 208.26
Chemical Name or Material Ethyl 4-ethoxyphenylacetate
Assay ≥97%
Molecular Formula C12H16O3
MDL Number MFCD00051917
CAS 40784-88-1
Beilstein 2969074
Synonym 4-Ethoxyphenylacetic acid ethyl ester
Boiling Point 112° to 113°C (0.5mmHg)
Quantity 5g