missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-ethoxybenzoate, 98%

Manufacturer:  Alfa Aesar™ A1501022

 View more versions of this product

Catalog No. AAA1501022



Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

Ethyl 4-ethoxybenzoate
145°C to 150°C (13mmHg)
100g
C11H14O3
2692236
HRAQMGWTPNOILP-UHFFFAOYSA-N
ethyl 4-ethoxybenzoate
90232
98%
137°C (278°F)
9°C
23676-09-7
MFCD00009116
benzoic acid, 4-ethoxy-, ethyl ester, 4-ethoxybenzoic acid ethyl ester, 4-ethoxy ethylbenzoate, benzoic acid, p-ethoxy-, ethyl ester, 4-ethoxyethylbenzoate, peeb, ethyl p-ethoxybenzoate, ethyl-4-ethoxybenzoate, p-ethoxy ethyl benzoate, ethyl-p-ethoxy benzoate
CCOC1=CC=C(C=C1)C(=O)OCC
194.23
194.23

Chemical Identifiers

23676-09-7
194.23
HRAQMGWTPNOILP-UHFFFAOYSA-N
90232
CCOC1=CC=C(C=C1)C(=O)OCC
C11H14O3
MFCD00009116
benzoic acid, 4-ethoxy-, ethyl ester, 4-ethoxybenzoic acid ethyl ester, 4-ethoxy ethylbenzoate, benzoic acid, p-ethoxy-, ethyl ester, 4-ethoxyethylbenzoate, peeb, ethyl p-ethoxybenzoate, ethyl-4-ethoxybenzoate, p-ethoxy ethyl benzoate, ethyl-p-ethoxy benzoate
ethyl 4-ethoxybenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents