Learn More
Ethyl 4-dimethylaminobenzoate, 99%
CAS: 10287-53-3 | C11H15NO2 | 193.246 g/mol
Supplier: Thermo Scientific Chemicals A1288822
Description
Ethyl 4-dimethylaminobenzoate is employed as a photoinititator in cell encapsulation applications. It is also useful for organic synthesis. Further, it is used in UV-curing coatings and inks for the initiation of photo polymerization of unsaturated prepolymers. In addition to this, it acts as a active pharmaceutical ingredient intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 4-dimethylaminobenzoate is employed as a photoinititator in cell encapsulation applications. It is also useful for organic synthesis. Further, it is used in UV-curing coatings and inks for the initiation of photo polymerization of unsaturated prepolymers. In addition to this, it acts as a active pharmaceutical ingredient intermediate.
Solubility
Insoluble in water.
Notes
Incompatible with reducing agents, oxidizing agents, bases and acids.
Specifications
| Ethyl 4-dimethylaminobenzoate | |
| 63°C to 65°C | |
| 190°C to 191°C (14 mmHg) | |
| MFCD00009115 | |
| 2210233 | |
| Insoluble in water. | |
| CCOC(=O)C1=CC=C(C=C1)N(C)C | |
| 193.246 | |
| CHEBI:52073 | |
| 99% |
| 10287-53-3 | |
| 1.06 | |
| C11H15NO2 | |
| 100 g | |
| ethyl 4-dimethylamino benzoate, parbenate, benzoic acid, 4-dimethylamino-, ethyl ester, kayacure epa, ethyl-p-dimethylaminobenzoate, ethyl-4-dimethylaminobenzoate, n,n-dimethylbenzocaine, speedcure edb, ethyl p-dimethylaminobenzoate, unii-829s8d3y0x | |
| FZUGPQWGEGAKET-UHFFFAOYSA-N | |
| ethyl 4-(dimethylamino)benzoate | |
| 25127 | |
| 193.25 |
Chemical Identifiers
| 10287-53-3 | |
| 193.246 | |
| FZUGPQWGEGAKET-UHFFFAOYSA-N | |
| 25127 | |
| ethyl 4-(dimethylamino)benzoate |
| C11H15NO2 | |
| MFCD00009115 | |
| ethyl 4-dimethylamino benzoate, parbenate, benzoic acid, 4-dimethylamino-, ethyl ester, kayacure epa, ethyl-p-dimethylaminobenzoate, ethyl-4-dimethylaminobenzoate, n,n-dimethylbenzocaine, speedcure edb, ethyl p-dimethylaminobenzoate, unii-829s8d3y0x | |
| CHEBI:52073 | |
| CCOC(=O)C1=CC=C(C=C1)N(C)C |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 233-634-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only