missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 4-Cyanobenzoate 98.0+%, TCI America™
$127.75 - $434.65
Chemical Identifiers
CAS | 7153-22-2 |
---|---|
Molecular Formula | C10H9NO2 |
Molecular Weight (g/mol) | 175.19 |
MDL Number | MFCD00001824 |
InChI Key | JLSSWDFCYXSLQX-UHFFFAOYSA-N |
Synonym | benzoic acid, 4-cyano-, ethyl ester, 4-cyanobenzoic acid ethyl ester, benzoic acid, 4-cyano-,ethyl ester, 4-cyano-benzoic acid ethyl ester, ethyl p-cyanobenzoate, ethyl-4-cyanobenzoate, p-cyano ethyl benzoate, acmc-209ypo, ethyl 4-cyanobenzoate, 4-cyanobenzoic acid, ethyl ester |
PubChem CID | 81589 |
IUPAC Name | ethyl 4-cyanobenzoate |
SMILES | CCOC(=O)C1=CC=C(C=C1)C#N |
Chemical Identifiers
7153-22-2 | |
175.19 | |
JLSSWDFCYXSLQX-UHFFFAOYSA-N | |
81589 | |
CCOC(=O)C1=CC=C(C=C1)C#N |
C10H9NO2 | |
MFCD00001824 | |
benzoic acid, 4-cyano-, ethyl ester, 4-cyanobenzoic acid ethyl ester, benzoic acid, 4-cyano-,ethyl ester, 4-cyano-benzoic acid ethyl ester, ethyl p-cyanobenzoate, ethyl-4-cyanobenzoate, p-cyano ethyl benzoate, acmc-209ypo, ethyl 4-cyanobenzoate, 4-cyanobenzoic acid, ethyl ester | |
ethyl 4-cyanobenzoate |
Specifications
7153-22-2 | |
White | |
C10H9NO2 | |
5 g | |
JLSSWDFCYXSLQX-UHFFFAOYSA-N | |
ethyl 4-cyanobenzoate | |
81589 | |
≥98.0% (GC) | |
Ethyl 4-Cyanobenzoate |
55°C | |
140°C | |
MFCD00001824 | |
benzoic acid, 4-cyano-, ethyl ester, 4-cyanobenzoic acid ethyl ester, benzoic acid, 4-cyano-,ethyl ester, 4-cyano-benzoic acid ethyl ester, ethyl p-cyanobenzoate, ethyl-4-cyanobenzoate, p-cyano ethyl benzoate, acmc-209ypo, ethyl 4-cyanobenzoate, 4-cyanobenzoic acid, ethyl ester | |
CCOC(=O)C1=CC=C(C=C1)C#N | |
175.19 | |
175.19 | |
Crystalline Powder |
Safety and Handling
TSCA : No