Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-cyano-5-(methylthio)thiophene-2-carboxylate, 97%

$161.23 - $403.81

Chemical Identifiers

Chemical Name or Material Ethyl 4-cyano-5-(methylthio)thiophene-2-carboxylate
CAS 116170-84-4
Molecular Formula C9H9NO2S2
Formula Weight 227.3
MDL Number MFCD01859680
Synonym 4-Cyano-5-(methylthio)thiophene-2-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2659403

  SDS
alfa aesar™
H2659403
1g Each for $403.81

AAH26594MD

  SDS
alfa aesar™
H26594MD
250mg Each for $161.23
Description & Specifications

Specifications

Formula Weight 227.3
Synonym 4-Cyano-5-(methylthio)thiophene-2-carboxylic acid ethyl ester
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Ethyl 4-cyano-5-(methylthio)thiophene-2-carboxylate
Quantity 1g
Melting Point 98° to 100°C
Molecular Formula C9H9NO2S2
Assay 97%
MDL Number MFCD01859680
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
CAS 116170-84-4
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.