Learn More
Ethyl 4-chlorophenylacetate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 388840250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl 4-chlorophenylacetate | |
Colorless | |
Authentic | |
Low Melting Solid | |
C10H11ClO2 | |
MFCD00018791 | |
UTWBWFXECVFDPZ-UHFFFAOYSA-N | |
ethyl 2-(4-chlorophenyl)acetate | |
84177 | |
98+% |
14062-24-9 | |
25 g | |
Glass bottle | |
98% min. (GC) | |
ClC6H4CH2C(O)OCH2CH3 | |
ethyl 4-chlorophenylacetate, ethyl 4-chlorophenyl acetate, ethyl 2-4-chlorophenyl acetate, 4-chlorophenyl acetic acid ethyl ester, ethyl p-chlorophenylacetate, benzeneacetic acid, 4-chloro-, ethyl ester, 4-chloro-phenyl-acetic acid ethyl ester, ethyl 4-chloro-phenylacetate, ksc522e5p | |
CCOC(=O)CC1=CC=C(C=C1)Cl | |
198.65 | |
198.65 |
Chemical Identifiers
14062-24-9 | |
UTWBWFXECVFDPZ-UHFFFAOYSA-N | |
84177 | |
CCOC(=O)CC1=CC=C(C=C1)Cl |
MFCD00018791 | |
ethyl 4-chlorophenylacetate, ethyl 4-chlorophenyl acetate, ethyl 2-4-chlorophenyl acetate, 4-chlorophenyl acetic acid ethyl ester, ethyl p-chlorophenylacetate, benzeneacetic acid, 4-chloro-, ethyl ester, 4-chloro-phenyl-acetic acid ethyl ester, ethyl 4-chloro-phenylacetate, ksc522e5p | |
ethyl 2-(4-chlorophenyl)acetate |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse
GHS Signal Word: Warning
EINECSNumber : 237-906-2
RUO – Research Use Only