missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-(Chloromethyl)benzoate, TCI America™

Manufacturer:  TCI America E066225G

 View more versions of this product

Catalog No. E066225G



Specifications

Specifications

Ethyl 4-(Chloromethyl)benzoate
Colorless
1201-90-7
1760
JTTXRFNOFFGPFI-UHFFFAOYSA-N
ethyl 4-(chloromethyl)benzoate
276519
Clear Liquid at 20°C
280°C
25g
C10H11ClO2
4-(Chloromethyl)benzoic Acid Ethyl Ester
CCOC(=O)C1=CC=C(C=C1)CCl
198.646
198.65
≥98.0% (GC)

Chemical Identifiers

1201-90-7
198.646
4-(Chloromethyl)benzoic Acid Ethyl Ester
ethyl 4-(chloromethyl)benzoate
C10H11ClO2
JTTXRFNOFFGPFI-UHFFFAOYSA-N
276519
CCOC(=O)C1=CC=C(C=C1)CCl
Safety and Handling

Safety and Handling

TSCA : No

Documents