Learn More
Ethyl 4-chloroacetoacetate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 177640500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethyl 4-chloroacetoacetate | |
638-07-3 | |
103.0°C (16.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4510 to 1.4530 | |
MFCD00000939 | |
03,663 | |
ethyl 4-chloroacetoacetate, 4-chloroacetoacetic acid ethyl ester, butanoic acid, 4-chloro-3-oxo-, ethyl ester, ethyl gamma-chloroacetoacetate, unii-5ae82250cm, ccris 6791, ethyl chloroacetyl acetate, butanoic acid, 4-chloro-3-oxo-ethylester, ethyl 4-chloro-3-oxobutyrate, ethyl 4-chloro-3-oxo-butanoate | |
OHLRLMWUFVDREV-UHFFFAOYSA-N | |
ethyl 4-chloro-3-oxobutanoate | |
69484 | |
98% |
98% | |
1.2100g/mL | |
96°C | |
97.5% min. (GC) | |
C6H9ClO3 | |
ClCH2COCH2CO2C2H5 | |
50g | |
1.21 | |
Solubility in water: 47.5g/L water (20°C). Other solubilities: soluble in ether,benzene,chloroform,,methylenechloride and carbontetrachloride | |
CCOC(=O)CC(=O)CCl | |
164.59 | |
164.59 |
Chemical Identifiers
638-07-3 | |
164.59 | |
OHLRLMWUFVDREV-UHFFFAOYSA-N | |
69484 | |
CCOC(=O)CC(=O)CCl |
C6H9ClO3 | |
MFCD00000939 | |
ethyl 4-chloroacetoacetate, 4-chloroacetoacetic acid ethyl ester, butanoic acid, 4-chloro-3-oxo-, ethyl ester, ethyl gamma-chloroacetoacetate, unii-5ae82250cm, ccris 6791, ethyl chloroacetyl acetate, butanoic acid, 4-chloro-3-oxo-ethylester, ethyl 4-chloro-3-oxobutyrate, ethyl 4-chloro-3-oxo-butanoate | |
ethyl 4-chloro-3-oxobutanoate |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specifi
GHS Signal Word: Danger
EINECSNumber : 211-317-0
RTECSNumber : AK5110000
TSCA : TSCA
RUO â Research Use Only