missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-chloroacetoacetate, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC177640010

 View more versions of this product

Catalog No. AC177640010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Ethyl 4-chloroacetoacetate
638-07-3
Authentic
1.4510 to 1.4530
98%
98%
Colorless to Red-Yellow
1kg
ClCH2COCH2CO2C2H5
Ethyl 4-chloro-3-oxobutanoate
CCOC(=O)CC(=O)CCl
164.585
164.59
98%
1.2100g/mL
Glass bottle
1.21
-8.0°C
103.0°C (16.0 mmHg)
C6H9ClO3
96°C
MFCD00000939
OHLRLMWUFVDREV-UHFFFAOYSA-N
ethyl 4-chloro-3-oxobutanoate
69484
Liquid

Chemical Identifiers

638-07-3
164.585
OHLRLMWUFVDREV-UHFFFAOYSA-N
69484
C6H9ClO3
MFCD00000939
Ethyl 4-chloro-3-oxobutanoate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 211-317-0

RTECSNumber : AK5110000

TSCA : TSCA

SDS Documents