missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-chloroacetoacetate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1564118

 View more versions of this product

Catalog No. AAA1564118



Description

Description

Ethyl 4-chloroacetoacetate is used as a precursor in the preparation of phosphorous ylides. It finds application in regular film coating, pharmaceutical coating and formulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-chloroacetoacetate is used as a precursor in the preparation of phosphorous ylides. It finds application in regular film coating, pharmaceutical coating and formulation.

Solubility
Miscible with water and most organic solvents.

Notes
Incompatible with strong acids, strong bases, reducing agents and strong oxidizing agents. Store in a cool place.
Specifications

Specifications

Ethyl 4-chloroacetoacetate
-8°C
220°C
C6H9ClO3
MFCD00000939
UN2922
ethyl 4-chloroacetoacetate, 4-chloroacetoacetic acid ethyl ester, butanoic acid, 4-chloro-3-oxo-, ethyl ester, ethyl gamma-chloroacetoacetate, unii-5ae82250cm, ccris 6791, ethyl chloroacetyl acetate, butanoic acid, 4-chloro-3-oxo-ethylester, ethyl 4-chloro-3-oxobutyrate, ethyl 4-chloro-3-oxo-butanoate
OHLRLMWUFVDREV-UHFFFAOYSA-N
ethyl 4-chloro-3-oxobutanoate
69484
97%
638-07-3
1.212
92°C (198°F)
1.452
50 g
1761275
Miscible with water and most organic solvents.
CCOC(=O)CC(=O)CCl
164.585
164.59

Chemical Identifiers

638-07-3
164.585
OHLRLMWUFVDREV-UHFFFAOYSA-N
69484
CCOC(=O)CC(=O)CCl
C6H9ClO3
MFCD00000939
ethyl 4-chloroacetoacetate, 4-chloroacetoacetic acid ethyl ester, butanoic acid, 4-chloro-3-oxo-, ethyl ester, ethyl gamma-chloroacetoacetate, unii-5ae82250cm, ccris 6791, ethyl chloroacetyl acetate, butanoic acid, 4-chloro-3-oxo-ethylester, ethyl 4-chloro-3-oxobutyrate, ethyl 4-chloro-3-oxo-butanoate
ethyl 4-chloro-3-oxobutanoate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301+H311-H314-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 211-317-0

RTECSNumber : AK5110000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only