missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-chloro-8-methylquinoline-3-carboxylate

$78.68 - $284.23

Chemical Identifiers

CAS 37041-32-0
Molecular Formula C13H12ClNO2
Formula Weight 249.7
MDL Number MFCD00173399
Synonym 4-Chloro-8-methylquinoline-3-carboxylic acid ethyl ester
Chemical Name or Material Ethyl 4-chloro-8-methylquinoline-3-carboxylate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5051003 SDS Alfa Aesar™
H5051003
1g Each for $284.23
AAH50510MD SDS Alfa Aesar™
H50510MD
250mg Each for $78.68
Specifications Description & Specifications

Chemical Identifiers

37041-32-0
249.7
4-Chloro-8-methylquinoline-3-carboxylic acid ethyl ester
C13H12ClNO2
MFCD00173399
Ethyl 4-chloro-8-methylquinoline-3-carboxylate

Specifications

37041-32-0
249.7
4-Chloro-8-methylquinoline-3-carboxylic acid ethyl ester
Ethyl 4-chloro-8-methylquinoline-3-carboxylate
C13H12ClNO2
MFCD00173399
1g