missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-chloro-8-fluoroquinoline-3-carboxylate, 97%

$126.27 - $436.81

Chemical Identifiers

CAS 56824-90-9
Molecular Formula C12H9ClFNO2
Formula Weight 253.66
MDL Number MFCD00173364
Synonym 4-Chloro-8-fluoroquinoline-3-carboxylic acid ethyl ester
Chemical Name or Material Ethyl 4-chloro-8-fluoroquinoline-3-carboxylate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5050703 SDS Alfa Aesar™
H5050703
1g Each for $436.81
AAH50507MD SDS Alfa Aesar™
H50507MD
250mg Each for $126.27
Specifications Description & Specifications

Chemical Identifiers

56824-90-9
253.66
4-Chloro-8-fluoroquinoline-3-carboxylic acid ethyl ester
C12H9ClFNO2
MFCD00173364
Ethyl 4-chloro-8-fluoroquinoline-3-carboxylate

Specifications

56824-90-9
253.66
4-Chloro-8-fluoroquinoline-3-carboxylic acid ethyl ester
Ethyl 4-chloro-8-fluoroquinoline-3-carboxylate
C12H9ClFNO2
MFCD00173364
1g
97%