Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate, 97%

$192.41 - $849.69

Chemical Identifiers

Chemical Name or Material Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate
CAS 77173-67-2
Molecular Formula C13H9ClN2O2
Formula Weight 260.68
MDL Number MFCD00173414
Synonym 4-Chloro-8-cyanoquinoline-3-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5051303

 
alfa aesar™
H5051303
1g Each for $192.41

AAH5051306

 
alfa aesar™
H5051306
5g Each for $849.69
Description & Specifications

Specifications

Formula Weight 260.68
Chemical Name or Material Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate
Assay 97%
Molecular Formula C13H9ClN2O2
MDL Number MFCD00173414
CAS 77173-67-2
Synonym 4-Chloro-8-cyanoquinoline-3-carboxylic acid ethyl ester
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.