Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-chloro-7,8-dimethylquinoline-3-carboxylate

$368.41 - $368.41

Chemical Identifiers

Chemical Name or Material Ethyl 4-chloro-7,8-dimethylquinoline-3-carboxylate
CAS 56824-88-5
Molecular Formula C14H14ClNO2
Formula Weight 263.73g/mol
MDL Number MFCD00665076
Synonym 4-Chloro-7,8-dimethylquinoline-3-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5052506

 
alfa aesar™
H50525-06
5g Each for $368.41
Description & Specifications

Specifications

Quantity 5g
Quantity 5g
Chemical Name or Material Ethyl 4-chloro-7,8-dimethylquinoline-3-carboxylate
CAS 56824-88-5
Molecular Formula C14H14ClNO2
Formula Weight 263.73g/mol
MDL Number MFCD00665076
Synonym 4-Chloro-7,8-dimethylquinoline-3-carboxylic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.