Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-chloro-6-methylquinoline-3-carboxylate

$163.96 - $163.96

Chemical Identifiers

Chemical Name or Material Ethyl 4-chloro-6-methylquinoline-3-carboxylate
CAS 56824-87-4
Molecular Formula C13H12ClNO2
Formula Weight 249.70g/mol
MDL Number MFCD00173357
Synonym 4-Chloro-6-methylquinoline-3-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5050503

 
alfa aesar™
H50505-03
Each for $163.96
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material Ethyl 4-chloro-6-methylquinoline-3-carboxylate
CAS 56824-87-4
Molecular Formula C13H12ClNO2
Formula Weight 249.70g/mol
MDL Number MFCD00173357
Synonym 4-Chloro-6-methylquinoline-3-carboxylic acid ethyl ester