Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate, 97%

$88.15 - $319.43

Chemical Identifiers

Chemical Name or Material Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate
CAS 22931-71-1
Molecular Formula C13H12ClNO3
Formula Weight 265.7
MDL Number MFCD00173417
Synonym 4-Chloro-6-methoxyquinoline-3-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5051503

  SDS
alfa aesar™
H5051503
1g Each for $319.43

AAH50515MD

  SDS
alfa aesar™
H50515MD
250mg Each for $88.15
Description & Specifications

Specifications

Formula Weight 265.7
MDL Number MFCD00173417
Molecular Formula C13H12ClNO3
Assay 97%
Synonym 4-Chloro-6-methoxyquinoline-3-carboxylic acid ethyl ester
CAS 22931-71-1
Quantity 1g
Melting Point 88° to 90°C
Chemical Name or Material Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate

Applications
Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.