Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-chloro-6-fluoroquinoline-3-carboxylate, 95%

$80.96 - $292.46

Chemical Identifiers

Chemical Name or Material Ethyl 4-chloro-6-fluoroquinoline-3-carboxylate
CAS 77779-49-8
Molecular Formula C12H9ClFNO2
Formula Weight 253.66
MDL Number MFCD00173350
Synonym 4-Chloro-6-fluoroquinoline-3-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5050203

  SDS
alfa aesar™
H5050203
1g Each for $292.46

AAH50502MD

  SDS
alfa aesar™
H50502MD
250mg Each for $80.96
Description & Specifications

Specifications

Formula Weight 253.66
Chemical Name or Material Ethyl 4-chloro-6-fluoroquinoline-3-carboxylate
Assay 95%
Molecular Formula C12H9ClFNO2
MDL Number MFCD00173350
CAS 77779-49-8
Synonym 4-Chloro-6-fluoroquinoline-3-carboxylic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.