Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-chloro-6,8-dimethylquinoline-3-carboxylate

$368.41 - $368.41

Chemical Identifiers

Chemical Name or Material Ethyl 4-chloro-6,8-dimethylquinoline-3-carboxylate
CAS 31602-09-2
Molecular Formula C14H14ClNO2
Formula Weight 263.73g/mol
MDL Number MFCD00173394
Beilstein 406421
Synonym 4-Chloro-6,8-dimethylquinoline-3-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5050806

 
alfa aesar™
H50508-06
5g Each for $368.41
Description & Specifications

Specifications

Quantity 5g
Quantity 5g
Chemical Name or Material Ethyl 4-chloro-6,8-dimethylquinoline-3-carboxylate
CAS 31602-09-2
Molecular Formula C14H14ClNO2
Formula Weight 263.73g/mol
MDL Number MFCD00173394
Beilstein 406421
Synonym 4-Chloro-6,8-dimethylquinoline-3-carboxylic acid ethyl ester