missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-chloro-6,8-difluoroquinoline-3-carboxylate

$170.54 - $741.97

Chemical Identifiers

CAS 150258-20-1
Molecular Formula C12H8ClF2NO2
Formula Weight 271.65
MDL Number MFCD00173353
Synonym 4-Chloro-6,8-difluoroquinoline-3-carboxylic acid ethyl ester
Chemical Name or Material Ethyl 4-chloro-6,8-difluoroquinoline-3-carboxylate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5050303 SDS Alfa Aesar™
H5050303
1g Each for $170.54
AAH5050306 SDS Alfa Aesar™
H5050306
5g Each for $741.97
Specifications

Description

Applications
Ethyl 4-chloro-6,8-difluoroquinoline-3-carboxylate is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Description & Specifications

Chemical Identifiers

150258-20-1
271.65
4-Chloro-6,8-difluoroquinoline-3-carboxylic acid ethyl ester
C12H8ClF2NO2
MFCD00173353
Ethyl 4-chloro-6,8-difluoroquinoline-3-carboxylate

Specifications

150258-20-1
271.65
4-Chloro-6,8-difluoroquinoline-3-carboxylic acid ethyl ester
Ethyl 4-chloro-6,8-difluoroquinoline-3-carboxylate
C12H8ClF2NO2
MFCD00173353
1g