Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-chloro-5,8-dimethylquinoline-3-carboxylate

$163.96 - $163.96

Chemical Identifiers

Chemical Name or Material Ethyl 4-chloro-5,8-dimethylquinoline-3-carboxylate
CAS 338954-51-1
Molecular Formula C14H14ClNO2
Formula Weight 263.73g/mol
MDL Number MFCD00173361
Synonym 4-Chloro-5,8-dimethylquinoline-3-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5050603

 
alfa aesar™
H50506-03
Each for $163.96
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material Ethyl 4-chloro-5,8-dimethylquinoline-3-carboxylate
CAS 338954-51-1
Molecular Formula C14H14ClNO2
Formula Weight 263.73g/mol
MDL Number MFCD00173361
Synonym 4-Chloro-5,8-dimethylquinoline-3-carboxylic acid ethyl ester