Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-chloro-5,7-dimethylquinoline-3-carboxylate

$170.27 - $747.35

Chemical Identifiers

Chemical Name or Material Ethyl 4-chloro-5,7-dimethylquinoline-3-carboxylate
CAS 338954-49-7
Molecular Formula C14H14ClNO2
Formula Weight 263.73
MDL Number MFCD00173354
Synonym 4-Chloro-5,7-dimethylquinoline-3-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5050403

  SDS
alfa aesar™
H5050403
1g Each for $170.27

AAH5050406

  SDS
alfa aesar™
H5050406
5g Each for $747.35
Description & Specifications

Specifications

Formula Weight 263.73
Chemical Name or Material Ethyl 4-chloro-5,7-dimethylquinoline-3-carboxylate
Molecular Formula C14H14ClNO2
MDL Number MFCD00173354
CAS 338954-49-7
Synonym 4-Chloro-5,7-dimethylquinoline-3-carboxylic acid ethyl ester
Quantity 1g