Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-chloro-5,7-difluoroquinoline-3-carboxylate, 99%

$119.45 - $412.53

Chemical Identifiers

Chemical Name or Material Ethyl 4-chloro-5,7-difluoroquinoline-3-carboxylate
CAS 311346-69-7
Molecular Formula C12H8ClF2NO2
Formula Weight 271.65
MDL Number MFCD00173416
Synonym 4-Chloro-5,7-difluorolquinoline-3-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5051403

  SDS
alfa aesar™
H5051403
1g Each for $412.53

AAH50514MD

  SDS
alfa aesar™
H50514MD
250mg Each for $119.45
Description & Specifications

Specifications

Formula Weight 271.65
Chemical Name or Material Ethyl 4-chloro-5,7-difluoroquinoline-3-carboxylate
Assay 99%
Molecular Formula C12H8ClF2NO2
MDL Number MFCD00173416
CAS 311346-69-7
Synonym 4-Chloro-5,7-difluorolquinoline-3-carboxylic acid ethyl ester
Quantity 1g