Please login to your online account to display your discounted pricing

Ethyl 4-chloro-3-nitrobenzoate, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  JP00516DA

 View more versions of this product

Catalog No. JP00516DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 229.62
Quantity 1g
Assay 97%
Chemical Name or Material Ethyl 4-chloro-3-nitrobenzoate
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C9H8ClNO4
CAS 16588-16-2

Specifications

Chemical Name or Material Ethyl 4-chloro-3-nitrobenzoate
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H8ClNO4
Formula Weight 229.62