Please login to your online account to display your discounted pricing

Ethyl 4-chloro-3-nitrobenzoate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Ethyl 4-chloro-3-nitrobenzoate
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H8ClNO4
Formula Weight 229.62
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

JP00516DA

  SDS
fisher scientific company
JP00516DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 229.62
Quantity 1g
Assay 97%
Chemical Name or Material Ethyl 4-chloro-3-nitrobenzoate
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C9H8ClNO4
CAS 16588-16-2