Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-chloro-3-nitrobenzoate, 97%

$53.49 - $53.49

Chemical Identifiers

Chemical Name or Material Ethyl 4-chloro-3-nitrobenzoate
CAS 16588-16-2
Molecular Formula C9H8ClNO4
Formula Weight 229.62
MDL Number MFCD00219621
Synonym 4-Chloro-3-nitrobenzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1933806

  SDS
alfa aesar™
A1933806
5g Each for $53.49
Description & Specifications

Specifications

Formula Weight 229.62
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
MDL Number MFCD00219621
Quantity 5g
Chemical Name or Material Ethyl 4-chloro-3-nitrobenzoate
CAS 16588-16-2
Synonym 4-Chloro-3-nitrobenzoic acid ethyl ester
Molecular Formula C9H8ClNO4
Melting Point 53° to 55°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.