Learn More
Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate, 98%
CAS: 5909-24-0 | C8H9ClN2O2S | 232.682 g/mol
Supplier: Thermo Scientific Chemicals B2242914
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate | |
| 60°C to 63°C | |
| MFCD00006085 | |
| ethyl 4-chloro-2-methylthio pyrimidine-5-carboxylate, ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, ethyl 4-chloro-2-methylthiopyrimidine-5-carboxylate, 4-chloro-2-methylsulfanyl-pyrimidine-5-carboxylic acid ethyl ester, ethyl 4-chloro-2-methylsulfanyl pyrimidine-5-carboxylate, 2-methylthio-4-chloro-5-ethoxycarbonylpyrimidine, 4-chloro-5-carbethoxy-2-methylthiopyrimidine, 5-pyrimidinecarboxylic acid, 4-chloro-2-methylthio-, ethyl ester, 4-chloro-2-methylthio pyrimidine-5-carboxylic acid ethyl ester, 4-chloro-2-methylsulfanylpyrimidine-5-carboxylic acid ethyl ester | |
| CCOC(=O)C1=CN=C(N=C1Cl)SC | |
| 232.682 | |
| 232.69 |
| 5909-24-0 | |
| C8H9ClN2O2S | |
| 25 g | |
| SNNHLSHDDGJVDM-UHFFFAOYSA-N | |
| ethyl 4-chloro-2-methylsulfanylpyrimidine-5-carboxylate | |
| 80008 | |
| 98% |
Chemical Identifiers
| 5909-24-0 | |
| 232.682 | |
| SNNHLSHDDGJVDM-UHFFFAOYSA-N | |
| 80008 | |
| CCOC(=O)C1=CN=C(N=C1Cl)SC |
| C8H9ClN2O2S | |
| MFCD00006085 | |
| ethyl 4-chloro-2-methylthio pyrimidine-5-carboxylate, ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, ethyl 4-chloro-2-methylthiopyrimidine-5-carboxylate, 4-chloro-2-methylsulfanyl-pyrimidine-5-carboxylic acid ethyl ester, ethyl 4-chloro-2-methylsulfanyl pyrimidine-5-carboxylate, 2-methylthio-4-chloro-5-ethoxycarbonylpyrimidine, 4-chloro-5-carbethoxy-2-methylthiopyrimidine, 5-pyrimidinecarboxylic acid, 4-chloro-2-methylthio-, ethyl ester, 4-chloro-2-methylthio pyrimidine-5-carboxylic acid ethyl ester, 4-chloro-2-methylsulfanylpyrimidine-5-carboxylic acid ethyl ester | |
| ethyl 4-chloro-2-methylsulfanylpyrimidine-5-carboxylate |
Safety and Handling
GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 227-619-0
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only