Learn More
Ethyl 4-bromocrotonate, tech. 80%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1037130
Description
Ethyl 4-bromocrotonate is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 4-bromocrotonate is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Insoluble in water.
Notes
Store at 4°C. Incompatible with acids, bases, oxidizing agents, reducing agents. Protect from heat. Store away from strong bases.
Specifications
Ethyl 4-bromocrotonate | |
1.402 | |
97°C (206°F) | |
1.493 | |
250 g | |
1721388 | |
Insoluble in water. | |
CCOC(=O)C=CCBr | |
193.04 | |
193.05 | |
Technical |
37746-78-4 | |
94°C to 95°C (12 mmHg) | |
C6H9BrO2 | |
MFCD00000247 | |
UN1760 | |
ethyl 4-bromocrotonate, e-ethyl 4-bromobut-2-enoate, ethyl 4-bromobut-2-enoate, 4-bromocrotonic acid ethyl ester, ethyl 2e-4-bromobut-2-enoate, 2-butenoic acid, 4-bromo-, ethyl ester, e, 2-butenoic acid, 4-bromo-, ethyl ester, ethyl e-4-bromo-2-butenoate, ethyl e-4-bromobut-2-enoate, ethyl trans-4-bromo-2-butenoate | |
FHGRPBSDPBRTLS-ONEGZZNKSA-N | |
ethyl (E)-4-bromobut-2-enoate | |
5373944 | |
80% |
Chemical Identifiers
37746-78-4 | |
193.04 | |
FHGRPBSDPBRTLS-ONEGZZNKSA-N | |
5373944 | |
CCOC(=O)C=CCBr |
C6H9BrO2 | |
MFCD00000247 | |
ethyl 4-bromocrotonate, e-ethyl 4-bromobut-2-enoate, ethyl 4-bromobut-2-enoate, 4-bromocrotonic acid ethyl ester, ethyl 2e-4-bromobut-2-enoate, 2-butenoic acid, 4-bromo-, ethyl ester, e, 2-butenoic acid, 4-bromo-, ethyl ester, ethyl e-4-bromo-2-butenoate, ethyl e-4-bromobut-2-enoate, ethyl trans-4-bromo-2-butenoate | |
ethyl (E)-4-bromobut-2-enoate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only