missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 4-bromocrotonate, 75%, tech., ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC118121000
Specifications
Ethyl 4-bromocrotonate | |
97°C | |
Glass bottle | |
1.4 | |
Yellow to Brown | |
tech., 75% | |
70% min. (GC) | |
BrCH2CH=CHCO2C2H5 | |
2-butenoic acid, 4-bromo-, ethyl ester, 2-butenoic acid, 4-bromo-, ethyl ester, e, 4-bromocrotonic acid ethyl ester, e-ethyl 4-bromobut-2-enoate, ethyl 2e-4-bromobut-2-enoate, ethyl 4-bromobut-2-enoate, ethyl 4-bromocrotonate, ethyl e-4-bromo-2-butenoate, ethyl e-4-bromobut-2-enoate, ethyl trans-4-bromo-2-butenoate | |
CCOC(=O)C=CCBr | |
193.04 | |
193.04 | |
75% |
1.4000g/mL | |
Authentic | |
1.4900 to 1.494 | |
94°C to 95°C (12.0mmHg) | |
100g | |
37746-78-4 | |
C6H9BrO2 | |
MFCD00000247 | |
FHGRPBSDPBRTLS-ONEGZZNKSA-N | |
ethyl (E)-4-bromobut-2-enoate | |
5373944 | |
Liquid | |
Technical |
Chemical Identifiers
37746-78-4 | |
193.04 | |
FHGRPBSDPBRTLS-ONEGZZNKSA-N | |
5373944 | |
CCOC(=O)C=CCBr |
C6H9BrO2 | |
MFCD00000247 | |
2-butenoic acid, 4-bromo-, ethyl ester, 2-butenoic acid, 4-bromo-, ethyl ester, e, 4-bromocrotonic acid ethyl ester, e-ethyl 4-bromobut-2-enoate, ethyl 2e-4-bromobut-2-enoate, ethyl 4-bromobut-2-enoate, ethyl 4-bromocrotonate, ethyl e-4-bromo-2-butenoate, ethyl e-4-bromobut-2-enoate, ethyl trans-4-bromo-2-butenoate | |
ethyl (E)-4-bromobut-2-enoate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
GHS Signal Word: Danger