Learn More
Ethyl 4-bromobutyrate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1347018
Description
Ethyl 4-bromobutyrate is used as a building block for the preparation of firefly luciferase inhibitor-conjucated peptide derivatives. It finds application in the synthesis of poly(2,6,-dimethyl-1,4-phehylene oxide) and thieno(2,3-b)-azepin-4-ones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 4-bromobutyrate is used as a building block for the preparation of firefly luciferase inhibitor-conjucated peptide derivatives. It finds application in the synthesis of poly(2,6,-dimethyl-1,4-phehylene oxide) and thieno(2,3-b)-azepin-4-ones.
Solubility
Immiscible with water.
Notes
Incompatible with bases, oxidizing agents and reducing agents.
Specifications
Ethyl 4-bromobutyrate | |
1.363 | |
90°C (194°F) | |
C6H11BrO2 | |
MFCD00000259 | |
1749700 | |
Immiscible with water. | |
CCOC(=O)CCCBr | |
195.056 | |
195.06 | |
Liquid |
2969-81-5 | |
80°C to 82°C (10 mm) | |
98% | |
1.456 | |
50 g | |
ethyl 4-bromobutyrate, 4-bromobutyric acid ethyl ester, ethyl-4-bromobutyrate, butanoic acid, 4-bromo-, ethyl ester, ethyl-4-bromotbutyrate, 4-bromo-n-butyric acid ethyl ester, brch2ch2ch2c o oc2h5, 4-bromo-butyric acid ethyl ester, 4-bromo-butanoic acid ethyl ester, gamma-bromobutyric acid ethyl ester | |
XBPOBCXHALHJFP-UHFFFAOYSA-N | |
ethyl 4-bromobutanoate | |
76300 | |
98% |
Chemical Identifiers
2969-81-5 | |
195.056 | |
XBPOBCXHALHJFP-UHFFFAOYSA-N | |
76300 | |
CCOC(=O)CCCBr |
C6H11BrO2 | |
MFCD00000259 | |
ethyl 4-bromobutyrate, 4-bromobutyric acid ethyl ester, ethyl-4-bromobutyrate, butanoic acid, 4-bromo-, ethyl ester, ethyl-4-bromotbutyrate, 4-bromo-n-butyric acid ethyl ester, brch2ch2ch2c o oc2h5, 4-bromo-butyric acid ethyl ester, 4-bromo-butanoic acid ethyl ester, gamma-bromobutyric acid ethyl ester | |
ethyl 4-bromobutanoate |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 221-005-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only