missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl (4-Bromobenzoyl)acetate, TCI America™

Manufacturer:  TCI America E11171G

 View more versions of this product

Catalog No. E11171G



Specifications

Specifications

Ethyl (4-Bromobenzoyl)acetate
Yellow
26510-95-2
MFCD00231243
PBDYXCKRDRCJDC-UHFFFAOYSA-N
ethyl 3-(4-bromophenyl)-3-oxopropanoate
2757149
Clear Liquid at 20°C
124°C
1g
C11H11BrO3
ethyl 3-4-bromophenyl-3-oxopropanoate, ethyl 4-bromobenzoyl acetate, ethyl 4-bromobenzoylacetate, 3-4-bromo-phenyl-3-oxo-propionic acid ethyl ester, 3-4-bromophenyl-3-oxo-propionic acid ethyl ester, ethyl 3-4-bromophenyl-3-oxo-propionate, benzenepropanoic acid, 4-bromo-beta-oxo-, ethyl ester, ethyl 4-bromobenzoyl acetate, ethyl p-bromobenzoyl acetate, benzenepropanoic acid,4-bromo-b-oxo-, ethyl ester
CCOC(=O)CC(=O)C1=CC=C(C=C1)Br
271.11
271.11
≥98.0% (GC)

Chemical Identifiers

26510-95-2
271.11
PBDYXCKRDRCJDC-UHFFFAOYSA-N
2757149
CCOC(=O)CC(=O)C1=CC=C(C=C1)Br
C11H11BrO3
MFCD00231243
ethyl 3-4-bromophenyl-3-oxopropanoate, ethyl 4-bromobenzoyl acetate, ethyl 4-bromobenzoylacetate, 3-4-bromo-phenyl-3-oxo-propionic acid ethyl ester, 3-4-bromophenyl-3-oxo-propionic acid ethyl ester, ethyl 3-4-bromophenyl-3-oxo-propionate, benzenepropanoic acid, 4-bromo-beta-oxo-, ethyl ester, ethyl 4-bromobenzoyl acetate, ethyl p-bromobenzoyl acetate, benzenepropanoic acid,4-bromo-b-oxo-, ethyl ester
ethyl 3-(4-bromophenyl)-3-oxopropanoate
Safety and Handling

Safety and Handling

TSCA : No

Documents