missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-Bromo-3-methylbenzoate, TCI America™

Manufacturer:  TCI America E103625G

 View more versions of this product

Catalog No. E103625G



Specifications

Specifications

Ethyl 4-Bromo-3-methylbenzoate
Yellow
160313-69-9
MFCD00673007
GIGDAWLJINYIFV-UHFFFAOYSA-N
ethyl 4-bromo-3-methylbenzoate
21729626
Clear Liquid at 20°C
275°C
25g
C10H11BrO2
4-Bromo-3-methylbenzoic Acid Ethyl Ester, Ethyl 4-Bromo-m-toluate, 4-Bromo-m-toluic Acid Ethyl Ester
CCOC(=O)C1=CC(=C(C=C1)Br)C
243.1
243.10
≥98.0% (GC)

Chemical Identifiers

160313-69-9
243.1
GIGDAWLJINYIFV-UHFFFAOYSA-N
21729626
CCOC(=O)C1=CC(=C(C=C1)Br)C
C10H11BrO2
MFCD00673007
4-Bromo-3-methylbenzoic Acid Ethyl Ester, Ethyl 4-Bromo-m-toluate, 4-Bromo-m-toluic Acid Ethyl Ester
ethyl 4-bromo-3-methylbenzoate
Safety and Handling

Safety and Handling

TSCA : No

Documents