Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-(benzyloxy)benzoate, 97%

$346.34 - $346.34

Chemical Identifiers

Chemical Name or Material Ethyl 4-(benzyloxy)benzoate
CAS 56441-55-5
Molecular Formula C16H16O3
Formula Weight 256.3
MDL Number MFCD00174341
Synonym 4-(Benzyloxy)benzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2068114

  SDS
alfa aesar™
B2068114
25g Each for $346.34
Description & Specifications

Specifications

Formula Weight 256.3
Chemical Name or Material Ethyl 4-(benzyloxy)benzoate
Assay 97%
Molecular Formula C16H16O3
MDL Number MFCD00174341
CAS 56441-55-5
Synonym 4-(Benzyloxy)benzoic acid ethyl ester
Melting Point 42° to 44°C
Quantity 25g