Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-aminopiperidine-1-carboxylate, 98%

$47.57 - $47.57

Chemical Identifiers

Chemical Name or Material Ethyl 4-aminopiperidine-1-carboxylate
CAS 58859-46-4
Molecular Formula C8H16N2O2
Formula Weight 172.23
MDL Number MFCD00006484
Beilstein 388597
Synonym 4-Aminopiperidine-1-carboxylic acid ethyl ester; 1-Ethoxycarbonyl-4-piperidinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1586606

 
alfa aesar™
L1586606
5g Each for $47.57
Description & Specifications

Specifications

Formula Weight 172.23
Chemical Name or Material Ethyl 4-aminopiperidine-1-carboxylate
Assay 98%
Molecular Formula C8H16N2O2
MDL Number MFCD00006484
CAS 58859-46-4
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 388597
Synonym 4-Aminopiperidine-1-carboxylic acid ethyl ester; 1-Ethoxycarbonyl-4-piperidinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Boiling Point 118° to 120°C (6mmHg)
Quantity 5g
Flash Point 78°C (172°F)
Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.