missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-aminocinnamate, 97%

$50.42 - $50.42

Chemical Identifiers

CAS 5048-82-8
Molecular Formula C11H13NO2
Formula Weight 191.23
MDL Number MFCD00017116
Beilstein 2803541
Synonym 4-Aminocinnamic acid ethyl ester
Chemical Name or Material Ethyl 4-aminocinnamate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0446403 SDS Alfa Aesar™
L0446403
1g Each for $50.42
Specifications Description & Specifications

Chemical Identifiers

5048-82-8
191.23
2803541
Ethyl 4-aminocinnamate
C11H13NO2
MFCD00017116
4-Aminocinnamic acid ethyl ester

Specifications

72° to 74°C
5048-82-8
191.23
2803541
Ethyl 4-aminocinnamate
1g
C11H13NO2
MFCD00017116
4-Aminocinnamic acid ethyl ester
97%

Safety and Handling

TSCA : TSCA