missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 4-Aminobutyrate Hydrochloride 98.0+%, TCI America™
Supplier: TCI America E092425G
Specifications
88°C | |
6937-16-2 | |
C6H14ClNO2 | |
ethyl 4-aminobutyrate hydrochloride, ethyl 4-aminobutanoate hydrochloride, 4-aminobutyric acid ethyl ester hydrochloride, ethyl 4-aminobutyrate hcl, butanoic acid, 4-amino-, ethyl ester, hydrochloride, 4-aminobutanoic acid ethyl ester hydrochloride, ethyl 4-aminobutanoate, chloride, 4-ethoxy-4-oxobutyl azanium chloride, h-gaba-oet.hcl, wln: z3vo2 &gh | |
CCOC(=O)CCCN.Cl | |
167.633 | |
167.63 | |
25 g |
Ethyl 4-Aminobutyrate Hydrochloride | |
White | |
MFCD00012915 | |
CXVQSUBJMYZELD-UHFFFAOYSA-N | |
ethyl 4-aminobutanoate;hydrochloride | |
12649661 | |
≥98.0% (N,T) | |
Crystalline Powder |
Chemical Identifiers
6937-16-2 | |
167.633 | |
CXVQSUBJMYZELD-UHFFFAOYSA-N | |
12649661 | |
CCOC(=O)CCCN.Cl |
C6H14ClNO2 | |
MFCD00012915 | |
ethyl 4-aminobutyrate hydrochloride, ethyl 4-aminobutanoate hydrochloride, 4-aminobutyric acid ethyl ester hydrochloride, ethyl 4-aminobutyrate hcl, butanoic acid, 4-amino-, ethyl ester, hydrochloride, 4-aminobutanoic acid ethyl ester hydrochloride, ethyl 4-aminobutanoate, chloride, 4-ethoxy-4-oxobutyl azanium chloride, h-gaba-oet.hcl, wln: z3vo2 &gh | |
ethyl 4-aminobutanoate;hydrochloride |
Safety and Handling
RTECSNumber : ES6650000
TSCA : No