Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-aminobutyrate hydrochloride, 98%

$35.27 - $97.19

Chemical Identifiers

Chemical Name or Material Ethyl 4-aminobutyrate hydrochloride
CAS 6937-16-2
Molecular Formula C6H13NO2·HCl
Linear Formula H2N(CH2)3CO2CH2CH3·HCl
Formula Weight 167.64
MDL Number MFCD00012915
Beilstein 3911780
Synonym 4-Aminobutyric acid ethyl ester hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0686306

  SDS
alfa aesar™
L0686306
5g Each for $35.27

AAL0686314

  SDS
alfa aesar™
L0686314
25g Each for $97.19
Description & Specifications

Specifications

Formula Weight 167.64
Chemical Name or Material Ethyl 4-aminobutyrate hydrochloride
CAS 6937-16-2
Beilstein 3911780
Synonym 4-Aminobutyric acid ethyl ester hydrochloride
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Molecular Formula C6H13NO2·HCl
Linear Formula H2N(CH2)3CO2CH2CH3·HCl
MDL Number MFCD00012915
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈70° to 75°C
Quantity 5g
RTECS Number ES6650000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.