Please login to your online account to display your discounted pricing

Ethyl 4-amino-1-piperidinecarboxylate, 98%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  100240050

 View more versions of this product

Catalog No. AC100240050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 172.23
Quantity 5mL
Color Clear colorless to light yellow
EINECS Number 261-472-3
Density 1.0040g/mL
Refractive Index 1.4810 to 1.4830 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 AIR SENSITIVE
Packaging Glass bottle
Synonym 4-Amino-1-(ethoxycarbonyl)piperidine
Infrared Spectrum Authentic
Water 0.5% max. (K.F.)
CAS 58859-46-4
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Name Note 98%
Assay Percent Range 98%
MDL Number MFCD00006484
Chemical Name or Material Ethyl 4-amino-1-piperidinecarboxylate
Molecular Formula C8H16N2O2

Specifications

Chemical Name or Material Ethyl 4-amino-1-piperidinecarboxylate
Name Note 98%
CAS 58859-46-4
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H16N2O2
Formula Weight 172.23
MDL Number MFCD00006484
Synonym 4-Amino-1-(ethoxycarbonyl)piperidine
Accessories

AIR SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

SDS & Safety Documents