missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-amino-1-piperidinecarboxylate, 98%, ACROS Organics™

Chemical Identifiers

CAS 58859-46-4
Assay Percent Range 98%
Molecular Formula C8H16N2O2
Formula Weight 172.23
MDL Number MFCD00006484
Synonym 4-Amino-1-(ethoxycarbonyl)piperidine
Chemical Name or Material Ethyl 4-amino-1-piperidinecarboxylate, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC100240250 SDS Thermo Fisher Scientific
100240250
Glass bottle 25mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC100240050 SDS Thermo Fisher Scientific
100240050
Glass bottle 5mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

58859-46-4
C8H16N2O2
MFCD00006484
Ethyl 4-amino-1-piperidinecarboxylate, 98%
98%
172.23
4-Amino-1-(ethoxycarbonyl)piperidine
≥97.5%

Specifications

58859-46-4
Authentic
1.4810 to 1.4830 (20°C, 589nm)
Liquid
Clear colorless to light yellow
25mL
MFCD00006484
Ethyl 4-amino-1-piperidinecarboxylate, 98%
1.0040g/mL
Glass bottle
0.5% max. (K.F.)
98%
C8H16N2O2
172.23
4-Amino-1-(ethoxycarbonyl)piperidine
≥97.5%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

AIR SENSITIVE

EINECSNumber : 261-472-3