missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-Acetylbenzoate, TCI America™

Manufacturer:  TCI America E11135G

 View more versions of this product

Catalog No. E11135G



Specifications

Specifications

Ethyl 4-Acetylbenzoate
54°C
38430-55-6
MFCD00013241
GLOAPLPTWAXAIG-UHFFFAOYSA-N
ethyl 4-acetylbenzoate
600911
Crystal-Powder at 20°C
White-Yellow
5g
C11H12O3
4-acetylbenzoic acid ethyl ester, benzoic acid, 4-acetyl-, ethyl ester, 4-acetyl-benzoic acid ethyl ester, ethyl4-acetylbenzoate, ethyl 4-acetylbenzate, pubchem18162, ethyl 4-acetyl benzoate, ethyl 4-acetyl-benzoate, acmc-1csm7, ethyl 4-acetylbenzoate
CCOC(=O)C1=CC=C(C=C1)C(=O)C
192.214
192.21
≥97.0% (GC)

Chemical Identifiers

38430-55-6
192.214
GLOAPLPTWAXAIG-UHFFFAOYSA-N
600911
CCOC(=O)C1=CC=C(C=C1)C(=O)C
C11H12O3
MFCD00013241
4-acetylbenzoic acid ethyl ester, benzoic acid, 4-acetyl-, ethyl ester, 4-acetyl-benzoic acid ethyl ester, ethyl4-acetylbenzoate, ethyl 4-acetylbenzate, pubchem18162, ethyl 4-acetyl benzoate, ethyl 4-acetyl-benzoate, acmc-1csm7, ethyl 4-acetylbenzoate
ethyl 4-acetylbenzoate
Safety and Handling

Safety and Handling

TSCA : No

Documents